Learn More
Medchemexpress LLC 3-MeOARh-NTR | 98.1% | 596.61 | 25 MG
SDP
Catalog No. 5000363993
Encompass_Preferred
3-MeOARh-NTR is an activatable imaging probe known for its high selectivity and good stability. It offers a high signal-to-noise ratio for nitroreductase (NTR) detection, making it an efficient molecular tool for sensing endogenous NTR. This probe is capable of producing a strong fluorescence signal in living HeLa cells under decreasing oxygen conditions and can facilitate fluorescence imaging of kidney tissues in mice, serving as an effective tool to evaluate kidney hypoxia.
  • High selectivity and good stability
  • High signal-to-noise ratio for nitroreductase (NTR) detection
  • Efficient molecular tool for endogenous NTR detection
  • Produces strong fluorescence signal in living HeLa cells with decreasing oxygen contents
  • Efficient probe to evaluate kidney hypoxia by NTR detection
Catalog No. 50-003-63993 Supplier Medchemexpress LLC Supplier No. 25MGHY149836
3-MeOARh-NTR is an activatable imaging probe known for its high selectivity and good stability. It offers a high signal-to-noise ratio for nitroreductase (NTR) detection, making it an efficient molecular tool for sensing endogenous NTR. This probe is capable of producing a strong fluorescence signal in living HeLa cells under decreasing oxygen conditions and can facilitate fluorescence imaging of kidney tissues in mice, serving as an effective tool to evaluate kidney hypoxia.
  • High selectivity and good stability
  • High signal-to-noise ratio for nitroreductase (NTR) detection
  • Efficient molecular tool for endogenous NTR detection
  • Produces strong fluorescence signal in living HeLa cells with decreasing oxygen contents
  • Efficient probe to evaluate kidney hypoxia by NTR detection