Learn More
Medchemexpress LLC 3-Mercapto-1,2,4-triazole | 3179-31-5 | 98.0% | 101.13 | 250 G
SDP
Catalog No. 5000451265
Encompass_Preferred
1,2-Dihydro-3H-1,2,4-triazole-3-thione is a biochemical reagent suitable for life science related research. It serves as a biological material or organic compound, intended for research use only and not for patient applications.
  • Utilized in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.
Catalog No. 50-004-51265 Supplier Medchemexpress LLC Supplier No. 250GHYW012991
1,2-Dihydro-3H-1,2,4-triazole-3-thione is a biochemical reagent suitable for life science related research. It serves as a biological material or organic compound, intended for research use only and not for patient applications.
  • Utilized in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.