Learn More
Medchemexpress LLC 3-Mercapto-1,2,4-triazole | 3179-31-5 | 99.8% | 101.13 | 50 G
SDP
Catalog No. 5000453443
Encompass_Preferred
1,2-Dihydro-3H-1,2,4-triazole-3-thione is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.
  • Biochemical reagent
  • Can be used as a biological material or organic compound for life science related research
Catalog No. 50-004-53443 Supplier Medchemexpress LLC Supplier No. HYW01299150G
1,2-Dihydro-3H-1,2,4-triazole-3-thione is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.
  • Biochemical reagent
  • Can be used as a biological material or organic compound for life science related research