Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercapto-1,2-propanediol, 90 wt% aqueous solution

Catalog No. AC125521000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125521000 100 mL Glass bottle
AC125520250 25 mL Glass bottle
AC125525000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC125521000 Supplier Thermo Scientific Chemicals Supplier No. 125521000
Only null left
Add to Cart

CAS: 96-27-5 | C3H8O2S | 108.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-27-5 , 7732-18-5
Molecular Formula C3H8O2S
Molecular Weight (g/mol) 108.16
MDL Number MFCD00004879
InChI Key PJUIMOJAAPLTRJ-UHFFFAOYNA-N
Synonym 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol
PubChem CID 7291
ChEBI CHEBI:74537
IUPAC Name 3-sulfanylpropane-1,2-diol
SMILES OCC(O)CS

Specifications

CAS Min % 88.0, 8.0
CAS Max % 92.0, 12.0
Color Colorless to Yellow
Density 1.2300g/mL
Boiling Point 118.0°C (5.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5073
Linear Formula HSCH2CH(OH)CH2OH
Quantity 100 mL
Beilstein 01,519
Merck Index 15,9488
Specific Gravity 1.23
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,insoluble in ether
Formula Weight 108.16
Percent Purity ≥88%
Physical Form Viscous Solution
Chemical Name or Material 3-Mercapto-1, 2-propanediol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get
EINECSNumber 202-495-0
RTECSNumber TY8140000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.