Learn More
Medchemexpress LLC 3-mercapto-1-propanol | 19721-22-3 | MFCD00192262 | 99.7% | 92.16 g/mol | C3H8OS | 25 G
SDP
Catalog No. 5000208841
Encompass_Preferred
3-mercapto-1-propanol is a thiol-alcohol reagent used in surface chemistry and synthetic applications. It is employed to prepare hydrophilic self-assembled monolayers and to act as a non-marking linker for chemical and enzymatic synthesis of oligosaccharides. The compound is a small, volatile liquid with CAS number 19721-22-3 and a molecular weight of 92.16 g/mol.
  • High purity: 99.7%.
  • Contains both a thiol and a primary alcohol functional group for surface coupling and derivatization.
  • Common applications include self-assembled monolayer formation and oligosaccharide linker chemistry.
  • Physical properties: molecular formula C3H8OS; molecular weight 92.16 g/mol.
  • Packaging available: 25 G, 50 G, 100 G, and larger quantities by request.
Catalog No. 50-002-08841 Supplier Medchemexpress LLC Supplier No. 25GHYW090210
3-mercapto-1-propanol is a thiol-alcohol reagent used in surface chemistry and synthetic applications. It is employed to prepare hydrophilic self-assembled monolayers and to act as a non-marking linker for chemical and enzymatic synthesis of oligosaccharides. The compound is a small, volatile liquid with CAS number 19721-22-3 and a molecular weight of 92.16 g/mol.
  • High purity: 99.7%.
  • Contains both a thiol and a primary alcohol functional group for surface coupling and derivatization.
  • Common applications include self-assembled monolayer formation and oligosaccharide linker chemistry.
  • Physical properties: molecular formula C3H8OS; molecular weight 92.16 g/mol.
  • Packaging available: 25 G, 50 G, 100 G, and larger quantities by request.