Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercapto-2-butanol (mixture of isomers) 97.0+%, TCI America™
SDP

Catalog No. M184325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M184325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M184325G Supplier TCI America Supplier No. M184325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54812-86-1
Molecular Formula C4H10OS
Molecular Weight (g/mol) 106.183
MDL Number MFCD00010271
InChI Key MJQWABQELVFQJL-UHFFFAOYSA-N
PubChem CID 62087
IUPAC Name 3-sulfanylbutan-2-ol
SMILES CC(C(C)S)O

Specifications

Color Yellow
Boiling Point 66°C
Quantity 25 g
Formula Weight 106.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Mercapto-2-butanol (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.