Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-mercaptoindole

Catalog No. 5000013321 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-000-13321 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-000-13321 Supplier Sigma Aldrich Supplier No. CDS014121250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,5-dimethoxytoluene (1,4-dimethoxy-2-methyl benzene) is a methoxy methyl benzene derivative
  • It has been synthesized from 2-methylhydroquinone
  • It is one of the volatile constituents identified from beechwood creosote and tuber brumale species
  • It is reported to be one of the thermochemolysis products in delaware bay particles and dissolved organic matters.2,5-dimethoxytoluene may be used as a reagent in the synthesis of 2,5-dimethoxy-4-methylbenzaldehyde-7-14c
  • Its IR spectrum has been used to establish the methyl group migration in the acid catalyzed rearrangement of 4-methoxy-4-methylcyclohexadienone

Specifications

CAS 480-94-4
Molecular Formula C8H7NS
Linear Formula C8H7NS
MDL Number MFCD00152110
Quantity 10 g
Molecular Weight (g/mol) 149.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.