Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercaptoisobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. M120025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M120025G 25 g
M12005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M120025G Supplier TCI America Supplier No. M120025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26473-47-2
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00671555
InChI Key MHRDCHHESNJQIS-UHFFFAOYSA-N
Synonym 3-Mercapto-2-methylpropionic Acid
PubChem CID 152910
IUPAC Name 2-methyl-3-sulfanylpropanoic acid
SMILES CC(CS)C(=O)O

Specifications

Color Yellow
Boiling Point 122°C
Quantity 25 g
Formula Weight 120.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Mercaptoisobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.