Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Mercaptopropanyl-N-hydroxysuccinimide ester

Catalog No. 9016995G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-169-95G 5 g
90-169-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-169-95G Supplier Sigma Aldrich Supplier No. 9016995G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Mercaptopropanyl-N-hydroxysuccinimide ester is an aliphatic, heterobifunctional crosslinking reagent. Due to the functional groups, this material can be used in a multitude of applications, such as the synthesis of protien-polymer conjugates and the synthesis of functionalized quantum dots. In addition to use as a crosslinking agent, this material can also be used to install functional groups post-polymerization. The terminal N-hydroxysuccinimide (NHS) ester can react with amines while the terminal thiol can react with thiols, alkenes, and alkynes. The free thiol can also be used to form self-assembled monolayers (SAMs) on gold surfaces.

Specifications

CAS 117235-10-6
Molecular Formula C7H9NO4S
Linear Formula C7H9NO4S
MDL Number MFCD28964889
Quantity 5 g
Synonym NHS ester
Molecular Weight (g/mol) 203.22
Percent Purity >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.