Learn More
Medchemexpress LLC HY-136159 500mg Medchemexpress, 3-Mercaptopropionic acid NHS ester CAS:117235-10-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY136159500MG
Encompass_Preferred
Medchemexpress, HY-136159 500mg 3-Mercaptopropionic acid NHS ester CAS:117235-10-6 3-Mercaptopropanyl-N-hydroxysuccinimide ester is an alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1425