Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-mercaptopropionic acid-d4 | 1141488-67-6 | 97.0% | 110.17 g/mol | C3H2D4O2S | 10 MG
SDP

Catalog No. 5000299373
Encompass_Preferred

3-Mercaptopropionic acid-d4 is the deuterium-labeled analogue of 3-mercaptopropionic acid used as a stable isotope reagent and analytical standard for research applications. It contains four deuterium atoms and is provided as a small-quantity vial for use in mass spectrometry, tracer studies, and method development.

  • Deuterium-labeled compound (d4) for isotope tracing and internal standard use.
  • High purity (97.0%) suitable for analytical applications.
  • Molecular weight 110.17 g/mol and formula C3H2D4O2S.
  • Supplied as a 10 mg small-quantity vial for laboratory use.
  • Viscous liquid, colorless to light yellow, easy to handle and store.

Catalog No. 50-002-99373 Supplier Medchemexpress LLC Supplier No. HYW032375S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-Mercaptopropionic acid-d4 is the deuterium-labeled analogue of 3-mercaptopropionic acid used as a stable isotope reagent and analytical standard for research applications. It contains four deuterium atoms and is provided as a small-quantity vial for use in mass spectrometry, tracer studies, and method development.

  • Deuterium-labeled compound (d4) for isotope tracing and internal standard use.
  • High purity (97.0%) suitable for analytical applications.
  • Molecular weight 110.17 g/mol and formula C3H2D4O2S.
  • Supplied as a 10 mg small-quantity vial for laboratory use.
  • Viscous liquid, colorless to light yellow, easy to handle and store.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.