Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-mercaptopropionic acid-d4 | 1141488-67-6 | 97.0% | 110.17 g/mol | C3H2D4O2S | 5 MG
SDP

Catalog No. 5000300205
Encompass_Preferred

3-Mercaptopropionic acid-d4 is the deuterium-labeled analogue of 3-mercaptopropionic acid, intended for use as a stable-isotope standard in analytical and research workflows. Supplied as a purified viscous liquid, the product is accompanied by documentation for safe handling and quality verification, and it has specified storage recommendations to preserve stability.

  • Deuterium-labeled analogue for isotope-dilution and mass spectrometry.
  • High purity (97.0%) for analytical reliability.
  • Molecular weight 110.17 g/mol and formula C3H2D4O2S.
  • CAS registry number 1141488-67-6 for unambiguous identification.
  • Available in small milligram quantities suitable for standards and controls.
  • Storage and handling guidance provided for long-term stability.
  • Safety data sheet (SDS) and certificate of analysis (COA) included for quality and safety.

Catalog No. 50-003-00205 Supplier Medchemexpress LLC Supplier No. HYW032375S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Mercaptopropionic acid-d4 is the deuterium-labeled analogue of 3-mercaptopropionic acid, intended for use as a stable-isotope standard in analytical and research workflows. Supplied as a purified viscous liquid, the product is accompanied by documentation for safe handling and quality verification, and it has specified storage recommendations to preserve stability.

  • Deuterium-labeled analogue for isotope-dilution and mass spectrometry.
  • High purity (97.0%) for analytical reliability.
  • Molecular weight 110.17 g/mol and formula C3H2D4O2S.
  • CAS registry number 1141488-67-6 for unambiguous identification.
  • Available in small milligram quantities suitable for standards and controls.
  • Storage and handling guidance provided for long-term stability.
  • Safety data sheet (SDS) and certificate of analysis (COA) included for quality and safety.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.