Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Mercaptopropyl(dimethoxy)methylsilane 95.0+%, TCI America™
SDP

Catalog No. M132325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M132325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M132325ML Supplier TCI America Supplier No. M132325ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 31001-77-1
Molecular Formula C6H16O2SSi
Molecular Weight (g/mol) 180.337
MDL Number MFCD00053923
InChI Key IKYAJDOSWUATPI-UHFFFAOYSA-N
Synonym 3-mercaptopropylmethyldimethoxysilane, 3-mercaptopropylmethyldimethoxy silane, 3-dimethoxymethylsilyl-1-propanethiol, 3-mercaptopropyl methyldimethoxysilane, dimethoxy-3-mercaptopropyl methylsilane, 3-dimethoxy methyl silyl propane-1-thiol, acmc-1aiak, ksc496q0j, 3-methyldimethoxysilyl propylmercaptan
PubChem CID 2759524
IUPAC Name 3-[dimethoxy(methyl)silyl]propane-1-thiol
SMILES CO[Si](C)(CCCS)OC

Specifications

Color Colorless
Boiling Point 74°C
Quantity 25 mL
Formula Weight 180.34
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Mercaptopropyl(dimethoxy)methylsilane
EINECSNumber (2)-3498
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.