Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Mercaptopropyl)Methyldimethoxysilane

Catalog No. 6379650ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-796-50ML 50 mL
63-796-250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 63-796-50ML Supplier Sigma Aldrich Supplier No. 6379650ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-Mercaptopropyl)methyldimethoxysilane (MPMDMS) can be used as a silane precursor in the synthesis of:Nafion-MPMDMS nanocomposite membranes by sol-gel method.MPMDMS functionalized magnetic- polysilsesquioxane (PSSQ) nanoparticles applicable for heavy metal ions adsorption.Thiol-organosilica nanoparticles.

Specifications

CAS 31001-77-1
Density 1 g/mL at 25°C
Boiling Point 96°C (30 mmHg, lit.)
Molecular Formula C6H16O2SSi
Refractive Index n20/D 1.45; n20/D 1.450
Linear Formula CH3Si(OCH3)2CH2CH2CH2SH
MDL Number MFCD00053923
Quantity 50 mL
Synonym Dimethoxy-(3-mercaptopropyl)methylsilane
Molecular Weight (g/mol) 180.34 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.