Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Mercaptopropyl)Triethoxysilane

Catalog No. 6379725MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-797-25MLF 25 mL
63-797-100MLF 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63-797-25MLF Supplier Sigma Aldrich Supplier No. 6379725MLF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-Mercaptopropyl)triethoxysilane (MPTS) can be used as a reagent to prepare thiol functionalized materials. Silica, SBA-15, alumina, starch, and graphene can be functionalized with MPTS and used in various applications.MPTS functionalized SBA-15 probe is used to determine dissolved mercury in solution.

Specifications

CAS 14814-09-6
Density 0.987 g/mL at 20°C
Molecular Formula C9H22O3SSi
Linear Formula HS(CH2)3Si(OCH2CH3)3
MDL Number MFCD00053924
Quantity 25 mL
Synonym 3-Triethoxysilyl-1-propanethiol
Molecular Weight (g/mol) 238.42 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.