Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Mercaptopropyl)trimethoxysilane, 85%, Tech., ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
4420-74-0
C6H16O3SSi
196.34
MFCD00004901
UUEWCQRISZBELL-UHFFFAOYSA-N
trimethoxysilylpropanethiol, 3-mercaptopropyltrimethoxysilane, 3-mercaptopropyl trimethoxysilane, 1-propanethiol, 3-trimethoxysilyl, silquest a 189, 3-trimethoxysilyl propanethiol, prosil 196, silane a 189, union carbide a-189, sila-ace s 810
20473
3-trimethoxysilylpropane-1-thiol
CO[Si](CCCS)(OC)OC
4420-74-0
C6H16O3SSi
196.34
MFCD00004901
UUEWCQRISZBELL-UHFFFAOYSA-N
trimethoxysilylpropanethiol, 3-mercaptopropyltrimethoxysilane, 3-mercaptopropyl trimethoxysilane, 1-propanethiol, 3-trimethoxysilyl, silquest a 189, 3-trimethoxysilyl propanethiol, prosil 196, silane a 189, union carbide a-189, sila-ace s 810
20473
3-trimethoxysilylpropane-1-thiol
CO[Si](CCCS)(OC)OC
Viscosity 2 mPa.s ( 20°C)
Specific Gravity 1.05
Boiling Point 215°C
Flash Point 88°C
Refractive Index 1.4410 to 1.443
Infrared Spectrum Authentic
Quantity 50g
Color Colorless to Yellow
Packaging Glass bottle
Density 1.0500g/mL
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 80% min. (GC)
Linear Formula HSCH2CH2CH2Si(OCH3)3
Beilstein 10, 148A
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in methanol,ethanol,isopropanol,,mineral spirits,acetone,benzene,toluene and,xylene
Formula Weight 196.34
Physical Form Liquid
Percent Purity 85%
Grade Technical
Chemical Name or Material (3-Mercaptopropyl)trimethoxysilane
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.
Harmful in contact with skin.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-588-5