Learn More
Sigma Aldrich [3-(Methacryloylamino)propyl]trimethylammonium chloride solution
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Specifications
Specifications
| CAS | 51410-72-1 |
| Density | 1.053 g/mL (at 25°C) |
| Molecular Formula | C10H21ClN2O |
| Refractive Index | n20/D 1.43 |
| Linear Formula | H2C=C(CH3)CONH(CH2)3 N(CH3)3 Cl |
| Quantity | 500 mL |
| MDL Number | MFCD00011812 |
| Synonym | Acrylamido buffer solution Pk. ≥12.0 |
| Molecular Weight (g/mol) | 220.74 |
| Biological Activity | Respiratory system |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.