Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methanesulfonyl-2-oxo-1-imidazolidinecarbonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. M194425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M194425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M194425G Supplier TCI America Supplier No. M194425G

missing translation for 'validMainframeMsg'

Specifications

Melting Point 180°C
CAS 41762-76-9
Color White-Yellow
Molecular Formula C5H7ClN2O4S
MDL Number MFCD00272360
UN Number 3261
Synonym 1-Chlorocarbonyl-3-methanesulfonyl-2-imidazolidinone, 1-Chloroformyl-3-methanesulfonyl-2-imidazolidinone
InChI Key ZWTPALHHEULAPI-UHFFFAOYSA-N
SMILES CS(=O)(=O)N1CCN(C1=O)C(=O)Cl
IUPAC Name 3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl chloride
Molecular Weight (g/mol) 226.631
PubChem CID 3016299
Formula Weight 226.63
Percent Purity ≥98.0% (T)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material 3-Methanesulfonyl-2-oxo-1-imidazolidinecarbonyl Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.