Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-methoxy-2-hydroxyphenyl boronic acid

Catalog No. CDS006533 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS006533 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS006533 Supplier Sigma Aldrich Supplier No. CDS00653350MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-thiophenemethylamine is a potential ligand replacement for poly(3-hexylthiophene)/cdse hybrid solar cells.reactant involved in synthesis of:
  • Triazole-linked-thiopene conjugates for use as a biomimetic model for studies of metal detoxification and oxidative stress involving metallothionein
  • Serotonin 5-ht1a receptor antagonists which have neuroprotective affects against ischemic cell damage
  • Imidazole- and piperonyl-containing thiadiazoles and pyrimidines for use as inducible oxide synthase dimerization inhibitors
  • Optoelectronic segmented polyurethanesreactant involved in:
  • Studies of organocatalyzed asymmetric reductive amination of ketones
  • Metal-free aerobic oxidative coupling of amines to imines

Specifications

Molecular Formula C7H9BO4
Linear Formula C7H9BO4
MDL Number MFCD08689552
Quantity 5 g
Molecular Weight (g/mol) 167.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.