Learn More
Sigma Aldrich 3-methoxy-2-methylbenzoic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- (r)-3,7-dimethyl-1-octene is an optically active alkyl-α-olefin
- Its copolymerization with styrene, 1-vinylnaphthalene or 2-methylstyrene by ziegler-natta catalysts has been reported
- Preparation of copolymers of racemic and optically active 3,7-dimethyl-1-octene with ethylene in the presence of ziegler-natta isospecific catalysts have been studied
- Polymerization of (r,s)-3,7-dimethyl-1-octene in the presence of heterogeneous or homogeneous isotactic specific catalysts involves highly stereospecific insertion of (r,s)-3,7-dimethyl-1-octene into the metal-ch3 bond
Specifications
Specifications
| Molecular Formula | C9H10O3 |
| Linear Formula | C9H10O3 |
| MDL Number | MFCD02094039 |
| Quantity | 1 mL |
| Molecular Weight (g/mol) | 166.17 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.