Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-2-nitrobenzaldehyde, 97%

Catalog No. AC172670050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC172670050 5 g
AC172670010 1 g
AC172670250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC172670050 Supplier Thermo Scientific Chemicals Supplier No. 172670050
Only null left
Add to Cart

CAS: 53055-05-3 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53055-05-3
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007135
InChI Key GDTUACILWWLIJF-UHFFFAOYSA-N
PubChem CID 98867
IUPAC Name 3-methoxy-2-nitrobenzaldehyde
SMILES COC1=CC=CC(C=O)=C1[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 97.0°C to 101.0°C
Color Beige to White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(NO2)CHO
Quantity 5 g
Beilstein 08, 62
Synonym benzaldehyde, 3-methoxy-2-nitro, 3-methoxy-2-nitro-benzaldehyde, 2-nitro-3-methoxybenzaldehyde, pubchem13093, acmc-20agz4, 2-nitro-3-methoxy benzaldehyde, benzaldehyde,3-methoxy-2-nitro, 3-methoxy-2-nitrobenzaldehyde
Formula Weight 181.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-2-nitrobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 258-332-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.