Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-2-nitrobenzoic Acid 99.0+%, TCI America™
SDP

Catalog No. M10361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10361G 1 g
M103610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10361G Supplier TCI America Supplier No. M10361G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4920-80-3
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00007143
InChI Key YMOMYSDAOXOCID-UHFFFAOYSA-N
Synonym 3-methoxy-2-nitrobenzoic acid, 2-nitro-3-methoxybenzoic acid, benzoic acid, 3-methoxy-2-nitro, 2-nitro-m-anisic acid, m-anisic acid, 2-nitro, 3-methoxy-2-nitrobenzoicacid, pubchem12905, acmc-1ao65, ksc238g2p
PubChem CID 78632
SMILES COC1=CC=CC(=C1[N+](=O)[O-])C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 197.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-2-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.