Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-2-nitrotoluene 98.0+%, TCI America™
SDP

Catalog No. M11965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11965G 5 g
M119625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11965G Supplier TCI America Supplier No. M11965G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5345-42-6
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00007179
InChI Key MGBRGNWARSQECY-UHFFFAOYSA-N
Synonym 3-methyl-2-nitroanisole, 3-methoxy-2-nitrotoluene, 2-nitro-3-methylanisole, benzene, 1-methoxy-3-methyl-2-nitro, pubchem12903, acmc-209l7h, anisole, 3-methyl-2-nitro, 3-methoxy-1-methyl-2-nitrobenzene, #
PubChem CID 79291
IUPAC Name 1-methoxy-3-methyl-2-nitrobenzene
SMILES COC1=CC=CC(C)=C1[N+]([O-])=O

Specifications

Melting Point 49°C
Color Yellow
Boiling Point 170°C
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-2-nitrotoluene
EINECSNumber (3)-0799
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.