Learn More
Sigma Aldrich 3-Methoxy-2-propoxybenzaldehyde
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 2,3-dimethylfuran is a methyl substituted furan
- The gas-phase rate constant determined by the relative rate method for the reaction of 2,3-dimethylfuran with no3 radical was found to be (5.83 +/- 0.46)x10-11 and with oh radical was (12.6 +/- 0.4)x10-11 (in units of cm3 molecule-1 s-1)
- It is reported to be formed by the cyclization of (z)-3-methylpent-2-en-4-yn-1-ol by various methods.2,3-dimethylfuran may be used in the synthesis of 2,9-dioxabicyclo[3.3.1]nonane
Specifications
Specifications
| Molecular Formula | C11H14O3 |
| Linear Formula | C11H14O3 |
| MDL Number | MFCD03422449 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 194.23 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.