Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Methoxy-3-methyl-1-butanol

Catalog No. 408069250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-806-9250ML 250 mL
40-806-91L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-806-9250ML Supplier Sigma Aldrich Supplier No. 408069250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-methoxy-3-methyl-1-butanol is widely used as solvent for paints, inks, and fragrances
  • Kinetics of the reaction of 3-methoxy-3-methyl-1-butanol with oh radicals using a relative rate method has been reported
  • It has been identified as oxidation products of dyes such as methyl yellow, methyl red and methyl orange.3-methoxy-3-methyl-1-butanol may be employed as raw material for the production of industrial detergents

Specifications

CAS 56539-66-3
Density 0.926 g/mL (at 20°C (literature))
Boiling Point 173°C to 175°C (lit.)
Molecular Formula C6H14O2
Refractive Index n20/D 1.428 (literature)
Linear Formula (CH3)2 C(OCH3)CH2CH2OH
MDL Number MFCD00044771
Quantity 250 mL
Molecular Weight (g/mol) 118.17
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.