Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxy-4-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Catalog No. AAH3388003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3388003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3388003 Supplier Thermo Scientific Chemicals Supplier No. H3388003

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 853367-87-0
Molecular Formula C9H8BrF3O
Molecular Weight (g/mol) 269.061
MDL Number MFCD18398596
InChI Key MUVUCILNTIWNQL-UHFFFAOYSA-N
Synonym 3-methoxy-4-trifluoromethyl benzyl bromide, 4-bromomethyl-2-methoxy-1-trifluoromethyl benzene
PubChem CID 66523605
IUPAC Name 4-(bromomethyl)-2-methoxy-1-(trifluoromethyl)benzene
SMILES COC1=C(C=CC(=C1)CBr)C(F)(F)F

Specifications

Quantity 1 g
UN Number UN3261
Formula Weight 269.06
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Methoxy-4-(trifluoromethyl)benzyl bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.