Learn More
Medchemexpress LLC 3-methoxy-5-(2-pyridin-2-ylethynyl)pyridine | 524924-76-3 | 98.1% | 210.23 g/mol | C13H10N2O | 25MG
SDP
Catalog No. 5000238483
Encompass_Preferred
Methoxy-PEPy (3-methoxy-5-(2-pyridin-2-ylethynyl)pyridine) is a potent, selective negative allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5) used for in vitro pharmacology and receptor-binding research. It is supplied as a solid and can be provided as a DMSO solution; reported batch purity is ~98.1% and molecular weight is 210.23 g/mol. Intended for research use only.
  • Potent mGlu5 antagonist (reported IC50 ≈ 1 nM).
  • High reported purity (~98.1%).
  • Molecular weight 210.23 g/mol; formula C13H10N2O.
  • Available in small milligram quantities and as a 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-38483 Supplier Medchemexpress LLC Supplier No. HY1251025MG
Methoxy-PEPy (3-methoxy-5-(2-pyridin-2-ylethynyl)pyridine) is a potent, selective negative allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5) used for in vitro pharmacology and receptor-binding research. It is supplied as a solid and can be provided as a DMSO solution; reported batch purity is ~98.1% and molecular weight is 210.23 g/mol. Intended for research use only.
  • Potent mGlu5 antagonist (reported IC50 ≈ 1 nM).
  • High reported purity (~98.1%).
  • Molecular weight 210.23 g/mol; formula C13H10N2O.
  • Available in small milligram quantities and as a 10 mM solution in DMSO.
  • Intended for research use only; not for human or clinical use.