Learn More
Medchemexpress LLC 3-Methoxy-9H-Carbazo 5mg
SDP
Catalog No. 5000205517
Encompass_Preferred
3-Methoxy-9H-Carbazole induces caspase-3 activities and the cellular generation of eactive oxygen species 3-Methoxy-9H-Carbazole inhibits cancer cell proliferation and induces apoptosis[1]
Catalog No. 50-002-05517 Supplier Medchemexpress LLC Supplier No. HYW1064565MG
3-Methoxy-9H-Carbazole induces caspase-3 activities and the cellular generation of eactive oxygen species 3-Methoxy-9H-Carbazole inhibits cancer cell proliferation and induces apoptosis[1]