Learn More
Medchemexpress LLC 3-methoxy-n-(1-methylpiperidin-4-yl)-4-[[4-[(3-oxo-1,2-dihydroinden-4-yl)oxy]-5-(trifluoromethyl)pyr | 1227948-02-8 | 98.6% | 555.55 | C28H28F3N5O4 | 100MG
SDP
Catalog No. 5000239376
Encompass_Preferred
BI-4464 is a selective, ATP-competitive inhibitor of focal adhesion kinase (PTK2/FAK) used as a research tool in biochemical and cellular studies and as a PTK2 ligand for PROTAC development. It shows potent enzymatic inhibition and is supplied with analytical documentation for preclinical research.
  • Highly selective ATP-competitive PTK2/FAK inhibitor.
  • Potent enzymatic activity (IC50 17 nM).
  • Suitable as a PTK2 ligand for PROTAC design.
  • Supplied with certificate of analysis and safety data sheet for quality assurance.
  • Available in multiple pack sizes for research use.
Catalog No. 50-002-39376 Supplier Medchemexpress LLC Supplier No. HY124625100MG
BI-4464 is a selective, ATP-competitive inhibitor of focal adhesion kinase (PTK2/FAK) used as a research tool in biochemical and cellular studies and as a PTK2 ligand for PROTAC development. It shows potent enzymatic inhibition and is supplied with analytical documentation for preclinical research.
  • Highly selective ATP-competitive PTK2/FAK inhibitor.
  • Potent enzymatic activity (IC50 17 nM).
  • Suitable as a PTK2 ligand for PROTAC design.
  • Supplied with certificate of analysis and safety data sheet for quality assurance.
  • Available in multiple pack sizes for research use.