Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-methoxy-N-(3-(1-methyl-1H-pyrazol-5-yl)-4-(2-morpholinoethoxy)phenyl)benzamide | 887936-68-7 | 98.9% | 436.50 g/mol | C24H28N4O4 | 100 MG
SDP

Catalog No. 5000242072
Encompass_Preferred

Temanogrel is a research-grade small molecule that acts as a highly selective antagonist of the serotonin 5-HT2A receptor (reported Ki = 4.9 nM). Supplied as a solid powder for in vitro and preclinical research, it is provided at high purity for biochemical and pharmacological studies.

  • Highly selective 5-HT2A receptor antagonist (Ki = 4.9 nM).
  • Intended for research use only; not for human therapeutic use.
  • High purity (98.94%) solid powder, white to light yellow.
  • Molecular formula C24H28N4O4; molecular weight 436.50 g/mol.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in small milligram pack sizes suitable for laboratory studies.

Catalog No. 50-002-42072 Supplier Medchemexpress LLC Supplier No. HY10560100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Temanogrel is a research-grade small molecule that acts as a highly selective antagonist of the serotonin 5-HT2A receptor (reported Ki = 4.9 nM). Supplied as a solid powder for in vitro and preclinical research, it is provided at high purity for biochemical and pharmacological studies.

  • Highly selective 5-HT2A receptor antagonist (Ki = 4.9 nM).
  • Intended for research use only; not for human therapeutic use.
  • High purity (98.94%) solid powder, white to light yellow.
  • Molecular formula C24H28N4O4; molecular weight 436.50 g/mol.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in small milligram pack sizes suitable for laboratory studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.