Learn More
Medchemexpress LLC 3-methoxy-N-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]-4-(2-morpholin-4-yl-2-oxoethoxy)benzamide | 781628-99-7 | 98.0% | 483.54 g/mol | C24H25N3O6S | 10MG
SDP
Catalog No. 5000237142
Encompass_Preferred
SBI-477 is a small-molecule chemical probe that stimulates insulin signaling by deactivating the transcription factor MondoA, resulting in reduced expression of insulin-pathway suppressors and coordinated inhibition of triacylglyceride synthesis while enhancing basal glucose uptake in human skeletal myocytes.
  • Deactivates MondoA to stimulate insulin signaling.
  • Reduces TXNIP and ARRDC4 expression.
  • Inhibits triacylglyceride synthesis and increases basal glucose uptake.
  • Provided as a solid powder, supplied in a 10 mg package.
  • High purity: 98.0%.
  • Recommended storage: powder at -20°C (long-term) or 4°C (shorter term).
Catalog No. 50-002-37142 Supplier Medchemexpress LLC Supplier No. HY12441810MG
SBI-477 is a small-molecule chemical probe that stimulates insulin signaling by deactivating the transcription factor MondoA, resulting in reduced expression of insulin-pathway suppressors and coordinated inhibition of triacylglyceride synthesis while enhancing basal glucose uptake in human skeletal myocytes.
  • Deactivates MondoA to stimulate insulin signaling.
  • Reduces TXNIP and ARRDC4 expression.
  • Inhibits triacylglyceride synthesis and increases basal glucose uptake.
  • Provided as a solid powder, supplied in a 10 mg package.
  • High purity: 98.0%.
  • Recommended storage: powder at -20°C (long-term) or 4°C (shorter term).