Learn More
Sigma Aldrich 3-methoxy-pyridine-2-carbaldehyde
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 3-methyl-2-thiophenecarboxaldehyde was used in the synthesis of 2,3-dimethyl-5-(2,6,10-trimethylundecyl)thiophene
- It was used to investigate versatile bioconversion capacity of bakerαs yeast for the generation of thiols from cysteine-aldehyde conjugates
Specifications
Specifications
| Molecular Formula | C7H7NO2 |
| Linear Formula | C7H7NO2 |
| MDL Number | MFCD05664031 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 137.14 |
| Biological Activity | Respiratory System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.