Learn More
Selleck Chemical LLC 3-Methoxybenzamide S5449-25mg
SDP
Catalog No. 503111528
Encompass_Preferred
3-Methoxybenzamide (3-MBA) is a competitive inhibitor of poly(ADP-ribose) synthetase with Ki values of less than 2 M and also inhibits ADP-ribosyltransferase (ADPRT)
Catalog No. 50-311-1528 Supplier Selleck Chemical LLC Supplier No. S544925MG
3-Methoxybenzamide (3-MBA) is a competitive inhibitor of poly(ADP-ribose) synthetase with Ki values of less than 2 M and also inhibits ADP-ribosyltransferase (ADPRT)