Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-methoxybenzamide | 5813-86-5 | 99.4% | 1 ML
SDP

Catalog No. 5000232425
Encompass_Preferred

3-methoxybenzamide is a small-molecule inhibitor of ADP-ribosyltransferases and poly(ADP-ribose) polymerase (PARP), supplied as a high-purity solid and as a ready-to-use 10 mM solution in DMSO for biochemical and cell-based assays.

  • High purity (99.4%) suitable for research applications.
  • Available as solid and as a 10 mM solution in DMSO (1 mL) for convenience.
  • Molecular formula C8H9NO2 and molecular weight 151.16 g/mol.
  • Commonly used as a PARP/ADPRT pathway modulator in biochemical studies.
  • Offered in multiple pack sizes to support small- and large-scale experiments.

Catalog No. 50-002-32425 Supplier Medchemexpress LLC Supplier No. HY12149710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-methoxybenzamide is a small-molecule inhibitor of ADP-ribosyltransferases and poly(ADP-ribose) polymerase (PARP), supplied as a high-purity solid and as a ready-to-use 10 mM solution in DMSO for biochemical and cell-based assays.

  • High purity (99.4%) suitable for research applications.
  • Available as solid and as a 10 mM solution in DMSO (1 mL) for convenience.
  • Molecular formula C8H9NO2 and molecular weight 151.16 g/mol.
  • Commonly used as a PARP/ADPRT pathway modulator in biochemical studies.
  • Offered in multiple pack sizes to support small- and large-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.