Learn More
Medchemexpress LLC 3-methoxybenzamide | 5813-86-5 | 99.4% | 10 MG
SDP
Catalog No. 5000231487
Encompass_Preferred
3-methoxybenzamide is a small-molecule inhibitor of ADP-ribosyltransferases (ADPRTs) and poly(ADP-ribose) polymerase (PARP) used as a research reagent and analytical standard. It affects bacterial septation via the FtsZ system and can enhance in vitro plant growth and microtuber formation.
  • Used as an inhibitor in cell cycle and DNA damage studies.
  • Affects bacterial cell division and septation.
  • Enhances plant tissue culture microtuber formation.
  • High purity (99.4%) suitable for analytical applications.
  • Molecular formula C8H9NO2; molecular weight 151.16.
Catalog No. 50-002-31487 Supplier Medchemexpress LLC Supplier No. HY12149710MG
3-methoxybenzamide is a small-molecule inhibitor of ADP-ribosyltransferases (ADPRTs) and poly(ADP-ribose) polymerase (PARP) used as a research reagent and analytical standard. It affects bacterial septation via the FtsZ system and can enhance in vitro plant growth and microtuber formation.
  • Used as an inhibitor in cell cycle and DNA damage studies.
  • Affects bacterial cell division and septation.
  • Enhances plant tissue culture microtuber formation.
  • High purity (99.4%) suitable for analytical applications.
  • Molecular formula C8H9NO2; molecular weight 151.16.