Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Methoxybenzamide | 5813-86-5 | 99.4% | 100 MG
SDP

Catalog No. 5000231195
Encompass_Preferred

3-Methoxybenzamide (3-MBA) is a small-molecule inhibitor of poly(ADP-ribose) polymerase (PARP) and ADP-ribosyltransferases (ADPRTs), supplied as a solid for research use. It is commonly used in biochemical and cell-based assays to study DNA repair pathways, PARP activity, and bacterial cell-division processes.

  • Small-molecule inhibitor of ADP-ribosyltransferase (ADPRT) and poly(ADP-ribose) polymerase (PARP).
  • Molecular formula C8H9NO2, molecular weight 151.16 g/mol.
  • High purity (99.4%) suitable for research applications.
  • Available as a solid and commonly supplied in 100 MG pack sizes.
  • Soluble in DMSO; 10 mM solution offered for convenience.
  • Used in biochemical assays and cell-based studies of DNA repair and cell cycle.

Catalog No. 50-002-31195 Supplier Medchemexpress LLC Supplier No. HY121497100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Methoxybenzamide (3-MBA) is a small-molecule inhibitor of poly(ADP-ribose) polymerase (PARP) and ADP-ribosyltransferases (ADPRTs), supplied as a solid for research use. It is commonly used in biochemical and cell-based assays to study DNA repair pathways, PARP activity, and bacterial cell-division processes.

  • Small-molecule inhibitor of ADP-ribosyltransferase (ADPRT) and poly(ADP-ribose) polymerase (PARP).
  • Molecular formula C8H9NO2, molecular weight 151.16 g/mol.
  • High purity (99.4%) suitable for research applications.
  • Available as a solid and commonly supplied in 100 MG pack sizes.
  • Soluble in DMSO; 10 mM solution offered for convenience.
  • Used in biochemical assays and cell-based studies of DNA repair and cell cycle.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.