Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-methoxybenzamide | 5813-86-5 | 99.4% | 200 MG
SDP

Catalog No. 5000233661
Encompass_Preferred

3-methoxybenzamide is a benzamide derivative used as a biochemical probe to inhibit ADP-ribosyltransferases (PARP) in research applications. It is a solid organic compound with formula C8H9NO2 and molecular weight 151.16 g/mol, typically used in biochemical assays and bacterial cell-division studies.

  • Inhibitor of ADP-ribosyltransferases (PARP) for biochemical and bacterial studies.
  • High purity solid suitable for analytical and research use.
  • Molecular formula: C8H9NO2.
  • Molecular weight: 151.16 g/mol.
  • Soluble in DMSO for preparation of concentrated stock solutions.

Catalog No. 50-002-33661 Supplier Medchemexpress LLC Supplier No. HY121497200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-methoxybenzamide is a benzamide derivative used as a biochemical probe to inhibit ADP-ribosyltransferases (PARP) in research applications. It is a solid organic compound with formula C8H9NO2 and molecular weight 151.16 g/mol, typically used in biochemical assays and bacterial cell-division studies.

  • Inhibitor of ADP-ribosyltransferases (PARP) for biochemical and bacterial studies.
  • High purity solid suitable for analytical and research use.
  • Molecular formula: C8H9NO2.
  • Molecular weight: 151.16 g/mol.
  • Soluble in DMSO for preparation of concentrated stock solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.