Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Methoxybenzamide | 5813-86-5 | 99.4% | 25 MG
SDP

Catalog No. 5000230225
Encompass_Preferred

3-methoxybenzamide is a small-molecule inhibitor of ADP-ribosyltransferases (ADPRTs) and poly(ADP-ribose) polymerases (PARPs), supplied as a high-purity solid for research use in biochemical assays and enzymatic studies.

  • High purity (99.4%).
  • Molecular weight 151.16 g/mol.
  • Molecular formula C8H9NO2.
  • Supplied as a solid powder with recommended cold storage.
  • Available in small research sizes including a 25 mg vial.

Catalog No. 50-002-30225 Supplier Medchemexpress LLC Supplier No. HY12149725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-methoxybenzamide is a small-molecule inhibitor of ADP-ribosyltransferases (ADPRTs) and poly(ADP-ribose) polymerases (PARPs), supplied as a high-purity solid for research use in biochemical assays and enzymatic studies.

  • High purity (99.4%).
  • Molecular weight 151.16 g/mol.
  • Molecular formula C8H9NO2.
  • Supplied as a solid powder with recommended cold storage.
  • Available in small research sizes including a 25 mg vial.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.