Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-methoxybenzamide | 5813-86-5 | 99.4% | 50 MG
SDP

Catalog No. 5000233888
Encompass_Preferred

3-methoxybenzamide is a small-molecule research reagent that inhibits ADP-ribosyltransferases (ADPRTs) and poly(ADP-ribose) polymerase (PARP). It is supplied as a solid, white to off-white powder and is used in bacterial and plant biology studies to probe cell division and tissue-culture responses.

  • Purity 99.4%
  • Solid, white to off-white powder
  • Molecular formula C8H9NO2
  • Molecular weight 151.16 g/mol
  • Reported activity: ADP-ribosyltransferase and PARP inhibitor
  • Available in small pack sizes including 50 MG

Catalog No. 50-002-33888 Supplier Medchemexpress LLC Supplier No. HY12149750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-methoxybenzamide is a small-molecule research reagent that inhibits ADP-ribosyltransferases (ADPRTs) and poly(ADP-ribose) polymerase (PARP). It is supplied as a solid, white to off-white powder and is used in bacterial and plant biology studies to probe cell division and tissue-culture responses.

  • Purity 99.4%
  • Solid, white to off-white powder
  • Molecular formula C8H9NO2
  • Molecular weight 151.16 g/mol
  • Reported activity: ADP-ribosyltransferase and PARP inhibitor
  • Available in small pack sizes including 50 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.