Learn More
Medchemexpress LLC 3-methoxybenzamide | 5813-86-5 | 99.4% | 500 MG
SDP
Catalog No. 5000231230
Encompass_Preferred
3-methoxybenzamide (3-MBA) is a small-molecule research compound that functions as an inhibitor of ADP-ribosyltransferases (ADPRTs) and PARP. It is provided as a solid for laboratory research and is commonly used in biochemical and cell-based studies.
  • High purity of 99.4%.
  • Molecular formula C8H9NO2 and molecular weight 151.16.
  • Available in milligram-scale packages for laboratory research.
  • Intended for research use only; not for human or animal therapeutic use.
Catalog No. 50-002-31230 Supplier Medchemexpress LLC Supplier No. HY121497500MG
3-methoxybenzamide (3-MBA) is a small-molecule research compound that functions as an inhibitor of ADP-ribosyltransferases (ADPRTs) and PARP. It is provided as a solid for laboratory research and is commonly used in biochemical and cell-based studies.
  • High purity of 99.4%.
  • Molecular formula C8H9NO2 and molecular weight 151.16.
  • Available in milligram-scale packages for laboratory research.
  • Intended for research use only; not for human or animal therapeutic use.