Learn More
Medchemexpress LLC 3-Methoxybenzeneboronic acid | 10365-98-7 | 151.96 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW000854
Encompass_Preferred
3-Methoxybenzeneboronic acid is a boronic acid derivative. It demonstrates a suitable binding pattern at the active site through non-covalent interactions with amino acid residues of proteins. It is a biochemical reagent used as a biological material or organic compound for life science related research.
  • Boronic acid derivative
  • Interacts non-covalently with amino acid residues of proteins
  • Functions as a biochemical reagent
  • Suitable for life science research
Catalog No. 50-004-49876