Learn More
Medchemexpress LLC 3-Methoxybenzeneboronic acid | 10365-98-7 | MFCD00161359 | 151.96 | 100 G
SDP
Catalog No. 5000449739
Encompass_Preferred
3-Methoxybenzeneboronic acid is a boronic acid derivative that exhibits a suitable binding pattern at the active site by non-covalently interacting with amino acid residues of proteins. It is a biochemical reagent used as a biological material or organic compound for life science-related research.
  • Boronic acid derivative
  • Exhibits suitable binding pattern at active site
  • Interacts non-covalently with amino acid residues of proteins
  • Biochemical reagent for life science-related research
  • Can be used as a biological material
  • Can be used as an organic compound
Catalog No. 50-004-49739 Supplier Medchemexpress LLC Supplier No. HYW000854100G
3-Methoxybenzeneboronic acid is a boronic acid derivative that exhibits a suitable binding pattern at the active site by non-covalently interacting with amino acid residues of proteins. It is a biochemical reagent used as a biological material or organic compound for life science-related research.
  • Boronic acid derivative
  • Exhibits suitable binding pattern at active site
  • Interacts non-covalently with amino acid residues of proteins
  • Biochemical reagent for life science-related research
  • Can be used as a biological material
  • Can be used as an organic compound