Learn More
Medchemexpress LLC 3-Methoxybenzeneboronic acid-d3 | 2241867-05-8 | 154.98 | 250 MG
SDP
Catalog No. 5000449955
Encompass_Preferred
This product, 3-Methoxybenzeneboronic acid-d3, is a deuterium labeled boronic acid derivative, also known as 3-Methoxyphenylboronic acid-d3. It is designed to interact non-covalently with amino acid residues of proteins at the active site, serving as a biochemical reagent for life science research.
  • Deuterium-labeled compound
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for analytical techniques
  • Soluble in DMSO
Catalog No. 50-004-49955 Supplier Medchemexpress LLC Supplier No. 250MGHYW000854S
This product, 3-Methoxybenzeneboronic acid-d3, is a deuterium labeled boronic acid derivative, also known as 3-Methoxyphenylboronic acid-d3. It is designed to interact non-covalently with amino acid residues of proteins at the active site, serving as a biochemical reagent for life science research.
  • Deuterium-labeled compound
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for analytical techniques
  • Soluble in DMSO