Learn More
Medchemexpress LLC 3-methoxybenzoic acid | 586-38-9 | MFCD00002499 | 99.6% | 152.15 g/mol | C8H8O3 | 50g
SDP
Catalog No. 5000268803
Encompass_Preferred
3-Methoxybenzoic acid can form complexes with metals such as europium (III) and gadolinium (III) 3-Methoxybenzoic acid can be metabolized by anaerobic bacteria through O-demethylation mechanism[1][2]
Catalog No. 50-002-68803 Supplier Medchemexpress LLC Supplier No. HYY076050G
Add to Cart
Add to Cart
3-Methoxybenzoic acid can form complexes with metals such as europium (III) and gadolinium (III) 3-Methoxybenzoic acid can be metabolized by anaerobic bacteria through O-demethylation mechanism[1][2]