Learn More
eMolecules​ 26905-40-8 | 3-Methoxybenzylmagnesium chloride, 0.25 M in 2-MeTHF | Synthonix | MFCD01319896 | 180.910 | C8H9ClMgO | 0.000 | [Cl-].COc1cccc(C[Mg+])c1 | 25ml | 627353681
Catalog No. 502695296 Shop All eMolecules Inc. Products
Encompass_Preferred
3-Methoxybenzylmagnesium chloride, 0.25 M in 2-MeTHF | Synthonix | 26905-40-8 | MFCD01319896 | 180.910 | C8H9ClMgO | 0.000 | [Cl-].COc1cccc(C[Mg+])c1 | 25ml | 627353681
Catalog No. 50-269-5296 Supplier eMolecules​ Supplier No. 627353681
3-Methoxybenzylmagnesium chloride, 0.25 M in 2-MeTHF | Synthonix | 26905-40-8 | MFCD01319896 | 180.910 | C8H9ClMgO | 0.000 | [Cl-].COc1cccc(C[Mg+])c1 | 25ml | 627353681