Learn More
Cayman Chemical 3-Methoxycatechol, Purity: ≥98%, CAS: 934-00-9, 10 g
SDP
Catalog No. 502912924
Encompass_Preferred
A lignan-derived phenol; a GPR35 agonist (EC50 = 147 µM in a DMR assay) using HT-29 cells; promotes esophageal carcinogenesis in rats at 2% in the diet either alone or in a multiorgan model of nitrosamine-induced carcinogenesis; has been used as a precursor in the enzymatic synthesis of pyrogallol and purpurogallin; has been used as a precursor in the electro-organic synthesis of coumestan derivatives
Catalog No. 50-291-2924 Supplier Cayman Chemical Supplier No. 3653510 G
A lignan-derived phenol; a GPR35 agonist (EC50 = 147 µM in a DMR assay) using HT-29 cells; promotes esophageal carcinogenesis in rats at 2% in the diet either alone or in a multiorgan model of nitrosamine-induced carcinogenesis; has been used as a precursor in the enzymatic synthesis of pyrogallol and purpurogallin; has been used as a precursor in the electro-organic synthesis of coumestan derivatives