Learn More
Cayman Chemical 3-Methoxycatechol 25g
SDP
Catalog No. 502912213
Encompass_Preferred
A lignan-derived phenol; a GPR35 agonist (EC50 = 147 µM in a DMR assay) using HT-29 cells; promotes esophageal carcinogenesis in rats at 2% in the diet either alone or in a multiorgan model of nitrosamine-induced carcinogenesis; has been used as a precursor in the enzymatic synthesis of pyrogallol and purpurogallin; has been used as a precursor in the electro-organic synthesis of coumestan derivatives
Catalog No. 50-291-2213 Supplier Cayman Chemical Supplier No. 3653525 G
A lignan-derived phenol; a GPR35 agonist (EC50 = 147 µM in a DMR assay) using HT-29 cells; promotes esophageal carcinogenesis in rats at 2% in the diet either alone or in a multiorgan model of nitrosamine-induced carcinogenesis; has been used as a precursor in the enzymatic synthesis of pyrogallol and purpurogallin; has been used as a precursor in the electro-organic synthesis of coumestan derivatives