Learn More
Medchemexpress LLC 3-Methoxycatechol 25g
SDP
Supplier:  Medchemexpress LLC 25GHYW116576
Encompass_Preferred
3-Methoxycatechol is intermediate for the antivascular agents combretastatin A-1 (HY-121993) and combretastatin B-1 3-Methoxycatechol promotes esophageal carcinogenesis in rats models[1][2]
Catalog No. 50-002-06845