Learn More
Medchemexpress LLC 3-Methoxycatechol 5g
SDP
Catalog No. 5000206679
Encompass_Preferred
3-Methoxycatechol is intermediate for the antivascular agents combretastatin A-1 (HY-121993) and combretastatin B-1 3-Methoxycatechol promotes esophageal carcinogenesis in rats models[1][2]
Catalog No. 50-002-06679 Supplier Medchemexpress LLC Supplier No. HYW1165765G
3-Methoxycatechol is intermediate for the antivascular agents combretastatin A-1 (HY-121993) and combretastatin B-1 3-Methoxycatechol promotes esophageal carcinogenesis in rats models[1][2]