Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxycatechol, 98%, Thermo Scientific™

Catalog No. AAB2000614
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2000614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2000614 Supplier Thermo Scientific Chemicals Supplier No. B2000614

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 934-00-9
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00002191
InChI Key LPYUENQFPVNPHY-UHFFFAOYSA-N
Synonym 3-methoxycatechol, 1,2-benzenediol, 3-methoxy, 3-methoxypyrocatechol, pyrocatechol, 3-methoxy, 2,3-dihydroxyanisole, pyrogallol 1-methyl ether, 1,2-dihydroxy-3-methoxybenzene, 6-methoxycatechol, pyrogallol 1-monomethyl ether, 3-methoxy-1,2-benzenediol
PubChem CID 13622
IUPAC Name 3-methoxybenzene-1,2-diol
SMILES COC1=CC=CC(=C1O)O

Specifications

Melting Point 40°C to 43°C
Boiling Point 146°C to 147°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1909165
Formula Weight 140.14
Percent Purity 98%
Chemical Name or Material 3-Methoxycatechol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-276-4
RTECSNumber CZ9002750
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.